missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 600 μS/cm at 25°C, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 99.96, 0.04 |
| CAS Max % | 99.96, 0.04 |
| Density | 1 |
| Physical Form | Liquid |
| Color | Colorless |
| pH | Approximately 7 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Solubility | Miscible with water |
| Show More |
Safety and Handling
| ShelfLife | 18 months |
| Recommended Storage | Ambient |