missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 500 μS/cm at 25°C, Ricca Chemical

Catalog No. p-3340515
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cube
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5887532 1 L Natural Poly Bottle
R58875004A 4 L Natural Poly Bottle
588755 20 L Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 5887532 Supplier Ricca Chemical Company Supplier No. 5887.532
Only null left

Potassium Chloride Conductivity Standard is tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.

  • Tested in ISO 17025 Accredited Laboratories.
  • Manufactured in cGMP compliant, FDA registered facilities.
  • Certified Traceable to NIST Standard Reference Material when available.
  • Many C of As and labels contain TDS and conductance information.
  • Tightest Specifications in the industry, lowest lot-to-lot variability.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0%, 0.03%
CAS Max % 100.0%, 0.03%
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Concentration or Composition (by Analyte or Components) 500 μS/cm (500 μmho/cm) at 25°C
Chemical Name or Material Potassium Chloride Conductivity Standard, Conductivity Standard
Show More Show Less
ShelfLife 12 months