missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 50 μS/cm at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 588651

 View more versions of this product

Catalog No. 13-622-606

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Chloride Conductivity Standard
Natural Poly Bottle
98.0,0.0025
Liquid
MFCD00011360
Approximately 7
Conductivity Standard
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4 L
CHEBI:32588
50μS/cm at 25°C
7447-40-7 , 7732-18-5
100.0,0.0026
ClK
Colorless
100°C
Conductivity Standard
[Cl-].[K+]
74.55
4873

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS
Documents

Documents

Promotions

Promotions