missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 50,000 μS/cm (50 mS/cm) at 25°C, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 95.0, 3.02 |
| CAS Max % | 99.0, 3.14 |
| Physical Form | Liquid |
| Color | Colorless |
| pH | Approximately 7 |
| Melting Point | Approximately 0°C |
| Solution Type | Conductivity Standard |
| Type | Conductivity Standard |
| Quantity | 4 L |
| Show More |