missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 5 μS/cm at 25°C, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Odor | Odorless |
| Assay Percent Range | 5.00 ± 0.03 μS/cm at 25°C |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Solubility | Infinitely soluble in water |
| Solution Type | Conductivity Standard |
| Type | Conductivity Standard |
| Solvent or Matrix | Water |
| Show More |