missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 5000 μS/cm at 25°C, Ricca Chemical

$152.17 - $696.72

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
58884316
View Documents
Ricca Chemical Company
58884316
500 mL Natural Poly Bottle N/A
Only null left
 
5888-4332
View Documents
Ricca Chemical Company
58884332
1 L Natural Poly Bottle N/A
Only null left
 
5888431
View Documents
Ricca Chemical Company
5888431
4 L Natural Poly Bottle N/A
Only null left
 
588843-5
View Documents
Ricca Chemical Company
5888435
20 L Cubitainer N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium chloride

Specifications

Natural Poly Bottle
98.0,0.27
Liquid
MFCD00011360
Approximately 7
Conductivity Standard
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
500 mL
CHEBI:32588
7447-40-7,7732-18-5
100.0,0.28
ClK
Colorless
100°C
Conductivity Standard
[Cl-].[K+]
74.55
4873
Potassium Chloride Conductivity Standard, 5 mS/cm (5000μmho/cm) at 25°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'