missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 5000 μS/cm at 25°C, Ricca Chemical

Catalog No. 5888435
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5888435 20 L Cubitainer
58884316 500 mL Natural Poly Bottle
58884332 1 L Natural Poly Bottle
5888431 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 5888435 Supplier Ricca Chemical Company Supplier No. 5888.435
Only null left

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Cubitainer
CAS Min % 98.0, 0.27
CAS Max % 100.0, 0.28
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 20 L
Chemical Name or Material Potassium Chloride Conductivity Standard, 5 mS/cm (5000μmho/cm) at 25°C
Show More Show Less