missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 4500 μS/cm at 25°C, Ricca Chemical

Catalog No. R58884201A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R58884201A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R58884201A Supplier Ricca Chemical Company Supplier No. R58884201A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.24
CAS Max % 100.0, 0.25
Physical Form Liquid
Color Colorless
pH Approximately 7
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Grade Laboratory
Chemical Name or Material Potassium Chloride Conductivity Standard
Show More Show Less