missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 3000 μS/cm at 25°C, Ricca Chemical

Catalog No. R58880404A
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R58880404A 4 L Natural Poly Bottle
58880432 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R58880404A Supplier Ricca Chemical Company Supplier No. R58880404A
Only null left

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 99.84, 0.16
CAS Max % 99.84, 0.16
Density 1.0011
Physical Form Liquid
Color Colorless
pH Approximately 7
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 4 L
Chemical Name or Material Potassium Chloride Conductivity Standard, 3000μS/cm at 25°C
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient