missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 3000 μS/cm at 25°C, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 98.0, 0.15 |
| CAS Max % | 100.0, 0.15 |
| Physical Form | Liquid |
| Color | Colorless |
| pH | Approximately 7 |
| Boiling Point | 100°C |
| Solution Type | Conductivity Standard |
| Type | Conductivity Standard |
| Quantity | 1 L |
| Show More |