missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 2500 μS/cm at 25°C, Ricca Chemical

Catalog No. p-4770371
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
Poly
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5888020500 500 mL Natural Poly Bottle
58880232 1 L Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R5888020500 Supplier Ricca Chemical Company Supplier No. R5888020500A
Only null left

Potassium Chloride Conductivity Standard is tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.

  • Tested in ISO 17025 Accredited Laboratories.
  • Manufactured in cGMP compliant, FDA registered facilities.
  • Certified Traceable to NIST Standard Reference Material when available.
  • Many C of As and labels contain TDS and conductance information.
  • Tightest Specifications in the industry, lowest lot-to-lot variability.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 2500 ± 10 μS/cm at 25°C
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solubility Infinitely soluble in water
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 500 mL
Chemical Name or Material Potassium Chloride Conductivity Standard, 2500 μS/cm at 25°C
Show More Show Less