missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 2500 μS/cm at 25°C, Ricca Chemical
Description
- Tested in ISO 17025 Accredited Laboratories.
- Manufactured in cGMP compliant, FDA registered facilities.
- Certified Traceable to NIST Standard Reference Material when available.
- Many C of As and labels contain TDS and conductance information.
- Tightest Specifications in the industry, lowest lot-to-lot variability.
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Odor | Odorless |
| Assay Percent Range | 2500 ± 10 μS/cm at 25°C |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Solubility | Infinitely soluble in water |
| Solution Type | Conductivity Standard |
| Type | Conductivity Standard |
| Solvent or Matrix | Water |
| Show More |