missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 250 μS/cm at 25°C, Ricca Chemical
Description
- Tested in our ISO 17025 Accredited Laboratories.
- Manufactured in cGMP compliant, FDA registered facilities.
- Certified Traceable to NIST Standard Reference Material when available.
- Many C of As and labels contain TDS and conductance information.
- Tightest Specifications in the industry, lowest lot-to-lot variability.
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 99.99%, 0.01% |
| CAS Max % | 99.99%, 0.01% |
| Density | 1 |
| Physical Form | Liquid |
| Color | Colorless |
| pH | Approximately 7 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Solubility | Miscible with water |
| Show More |
Safety and Handling
| ShelfLife | 24 months |
| Recommended Storage | Ambient |