missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 25 μS/cm at 25°C, Ricca Chemical

$311.82 - $311.82

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R58852504A
View Documents
Ricca Chemical Company
R58852504A
4 L Natural Poly Bottle
Each for $311.82
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium chloride

Specifications

Natural Poly Bottle
Odorless
Liquid
MFCD00011360
Approximately 7
Infinitely soluble in water
Conductivity Standard
[Cl-].[K+]
potassium chloride
4 L
CHEBI:32588
7447-40-7 , 7732-18-5
25.0 ± 0.1 uS/cm at 25°C
ClK
Colorless
100°C
Conductivity Standard
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Water
74.55
4873
Potassium Chloride Conductivity Standard, 25μS/cm (25μmho/cm) at 25°C
Videos
SDS
Documents

Documents