missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 25,000 μS/cm (25 mS/cm) at 25°C, Ricca Chemical

Catalog No. R5888600500
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
Poly
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5888600500 500 mL Natural Poly Bottle
5888632 1 L Poly
R588860010F 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R5888600500 Supplier Ricca Chemical Company Supplier No. R5888600500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Chloride Conductivity Standard is tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.

  • Tested in ISO 17025 Accredited Laboratories.
  • Manufactured in cGMP compliant, FDA registered facilities.
  • Certified Traceable to NIST Standard Reference Material when available.
  • Many C of As and labels contain TDS and conductance information.
  • Tightest Specifications in the industry, lowest lot-to-lot variability.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 25,000 ± 100 μS/cm at 25°C
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 500 mL
Chemical Name or Material Potassium Chloride Conductivity Standard, 25,000 μS/cm (25,000 μmho/cm) at 25°C
Show More Show Less