missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 20 μS/cm at 25°C, Ricca Chemical

Catalog No. R5885020500
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R5885020500 500 mL
R58850204A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R5885020500 Supplier Ricca Chemical Company Supplier No. R5885020500A
Only null left

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 100, 0.0011
CAS Max % 100, 0.0011
Density 0.9998
Physical Form Liquid
Color Colorless
pH Approximately 7
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Chemical Name or Material Potassium Chloride Conductivity Standard, 20μS/cm at 25°C
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient