missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 20,000 μS/cm (20 mS/cm) at 25°C, Ricca Chemical

Catalog No. R58882004A
Change view
Click to view available options
Quantity:
4 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R58882004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R58882004A Supplier Ricca Chemical Company Supplier No. R58882004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 20.0 ± 0.1 mS/cm at 25°C
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 4 L
Chemical Name or Material Potassium Chloride Conductivity Standard, 20 mS/cm (20, 000μmho/cm) at 25°C
Show More Show Less