missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 1800 μS/cm at 25°C

Catalog No. 588818032
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
588818032 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 588818032 Supplier Ricca Chemical Company Supplier No. R58881801A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Chloride Conductivity Standard is tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.

  • Tested in ISO 17025 Accredited Laboratories.
  • Manufactured in cGMP compliant, FDA registered facilities.
  • Certified Traceable to NIST Standard Reference Material when available.
  • Many C of As and labels contain TDS and conductance information.
  • Tightest Specifications in the industry, lowest lot-to-lot variability.

Chemical Identifiers

CAS 7732-18-5 , 7447-40-7

Specifications

Packaging Poly
Certificate of Analysis Yes
Traceability to NIST SRM# 999
CAS Min % 99.91%, 0.09%
CAS Max % 0.09%, 99.91%
Conductivity 1790 to 1810 μS/cm
Technique Calibration of Conductivity Meter
Certifications/Compliance ISO 17025
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Conductivity Meters and Electrodes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Grade Laboratory
Chemical Name or Material KCl Conductivity Standard
Show More Show Less
ShelfLife 24 Months