missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 100 μS/cm at 25°C, Ricca Chemical

Catalog No. R588700010F
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R588700010F 10 L Cubitainer
588716 500 mL Natural Poly Bottle
588732 1 L Natural Poly Bottle
13622604 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R588700010F Supplier Ricca Chemical Company Supplier No. R588700010F
Only null left

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Cubitainer
Odor Odorless
Assay Percent Range 100.0 ± 0.1 μS/cm at 25°C
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solubility Infinitely soluble in water
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 10 L
Chemical Name or Material Potassium Chloride Conductivity Standard, 100 μS/cm (100 μmho/cm) at 25°C
Show More Show Less