missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 100,000 μS/cm (100 mS/cm) at 25°C, Ricca Chemical

Catalog No. R5889300500
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5889300500 500 mL Natural Poly Bottle
R58893001A 1 L Natural Poly Bottle
R58893004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R5889300500 Supplier Ricca Chemical Company Supplier No. R5889300500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.0047
CAS Max % 100.0, 0.0049
Physical Form Liquid
Color Colorless
pH Approximately 7
Melting Point Approximately 0°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Chemical Name or Material Potassium Chloride Conductivity Standard, 100mS/cm (100,000μmho/cm) at 25°C
Show More Show Less