missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 10 μS/cm at 25°C, Ricca Chemical

Catalog No. 5885116
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5885116 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5885116 Supplier Ricca Chemical Company Supplier No. R5885100500A
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 10.00 ± 0.03 μS/cm at 25°C
Physical Form Liquid
Color Colorless
pH Approximately 7
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Infinitely soluble in water
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Potassium Chloride Conductivity Standard, 10μS/cm (10μmho/cm) at 25°C
Show More Show Less