missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C, Ricca Chemical

Catalog No. 58881016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58881016 500 mL
58881032 1 L
5888101 4 L
R588810010F 10 L
R588810020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 58881016 Supplier Ricca Chemical Company Supplier No. 5888.1016
Only null left

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.0, 0.56
CAS Max % 100.0, 0.59
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Chemical Name or Material Potassium Chloride Conductivity Standard, 10 mS/cm (10, 000μmho/cm) at 25°C
Show More Show Less