missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 1000 μS/cm at 25°C, Ricca Chemical

Catalog No. 5888011
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cube
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5888011 4 L Natural Poly Bottle
58880116 500 mL Natural Poly Bottle
58880132 1 L Natural Poly Bottle
R588801010F 10 L Cubitainer
5888015 20 L Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 5888011 Supplier Ricca Chemical Company Supplier No. 5888.011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.

  • Tested in ISO 17025 Accredited Laboratories.
  • Manufactured in cGMP compliant, FDA registered facilities.
  • Certified Traceable to NIST Standard Reference Material when available.
  • Many C of As and labels contain TDS and conductance information.
  • Tightest Specifications in the industry, lowest lot-to-lot variability.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0%, 0.05%
CAS Max % 100.0%, 0.05%
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 4 L
Chemical Name or Material Potassium Chloride Conductivity Standard, 1 mS/cm (1000μmho/cm) at 25°C
Show More Show Less