missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 1500 μS/cm at 25°C, Ricca Chemical

Catalog No. p-4770362
Change view
Click to view available options
Quantity:
1 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R58881501A 1 L Natural Poly Bottle
R588815010F 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R58881501A Supplier Ricca Chemical Company Supplier No. R58881501A
Only null left

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 1, 500 ± 1 μS/cm at 25°C
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 1 L
Chemical Name or Material Potassium Chloride Conductivity Standard, 1.5 mS/cm (1, 500μmho/cm) at 25°C
Show More Show Less