missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 1M aq. soln.

CAS: 7447-40-7 | ClK | 74.55 g/mol

Supplier:  Thermo Scientific Chemicals J63739AK

 View more versions of this product

Catalog No. AAJ63739AK


Only null left
Explore available promotions

Description

Description

Potassium chloride 1M solution is used as conductive standard and as electrolyte in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4873
74.55
Liquid
7447-40-7
KCl
1M aq. soln.
Miscible with water.
[Cl-].[K+]
74.55
CHEBI:32588
250 mL

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only