missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, 1M aq. soln.
CAS: 7447-40-7 | ClK | 74.55 g/mol
$154.63 - $287.74
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63739AE
|
Thermo Scientific Chemicals
J63739AE |
100 mL |
Each for $154.63
|
|
|||||
|
AAJ63739AK
|
Thermo Scientific Chemicals
J63739AK |
250 mL |
Each for $287.74
|
|
|||||
Description
Potassium chloride 1M solution is used as conductive standard and as electrolyte in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| potassium chloride |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 | |
| [Cl-].[K+] |
Specifications
| 7447-40-7 | |
| KCl | |
| 1M aq. soln. | |
| Miscible with water. | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| 100 mL | |
| Potassium chloride |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| 74.55 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only