missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 0.010M, Conductivity Standard

CAS: 7447-40-7 | ClK | 74.55 g/mol

Supplier:  Thermo Scientific Chemicals 042679K2

 View more versions of this product

Catalog No. AA42679K2


Only null left
Explore available promotions

Description

Description

Potassium chloride 0.010M solution is used as conductive standard and as electrolyte in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium chloride
7447-40-7 , 7732-18-5
23 hPa (17mm Hg) at 20°C
0.010M
ClK
Colorless
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
1 L
CHEBI:32588
Potassium chloride: 0.075%; Water: 99.925%
1413μmho at 25°C
Odorless
1g/cm3 at 20°C
Liquid
MFCD00011360
Miscible with water
[Cl-].[K+]
74.55
4873
74.55

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
ClK
MFCD00011360
4873
[Cl-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-211-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only