missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 0.00068M, Conductivity Standard

Catalog No. p-4782069
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43445AP 500 mL
AA43445K2 1 L
AA43445K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43445AP Supplier Thermo Scientific Chemicals Supplier No. 043445AP
Only null left

CAS: 7447-40-7 | ClK | 74.55 g/mol

Potassium chloride 0.00068M solution is used as conductive standard and as electrolyte in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Odor Odorless
Vapor Pressure 17mm Hg at 20°C
Assay Percent Range 0.00068M
Physical Form Liquid
Color Colorless
Solubility Miscible with water
IUPAC Name potassium chloride
Quantity 500 mL
Formula Weight 74.55
Concentration or Composition (by Analyte or Components) Potassium chloride: 0.005%; Water: 99.995%
Chemical Name or Material Potassium chloride
Show More Show Less
EINECSNumber 231-211-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only