missing translation for 'onlineSavingsMsg'
Learn More

Potassium chlorate, 99+%, ACS reagent

Catalog No. AC418190050
Change view
Click to view available options
Quantity:
5 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418190050 5 g Glass Bottle
AC418190025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418190050 Supplier Thermo Scientific Chemicals Supplier No. 418190050

CAS: 3811-04-9 | ClKO3 | 122.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
MDL Number MFCD00011361
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
PubChem CID 6426889
IUPAC Name potassium;chlorate
SMILES [O-]Cl(=O)=O.[K+]

Specifications

Melting Point 356.0°C to 368.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99+%
Linear Formula KClO3
Fieser 01,927
Merck Index 15, 7741
Solubility Information Solubility in water: 73g/L (20°C). Other solubilities: 555g/L hot water (100°C), soluble in alkalies, 1g/50mL glycerol, practically insoluble in alcohol, insoluble in ether
Formula Weight 122.55
Percent Purity ≥99%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Potassium chlorate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the ris
EINECSNumber 223-289-7
RTECSNumber FO0350000
TSCA TSCA

RUO – Research Use Only