missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, EMSURE™ MilliporeSigma™

Catalog No. M1049280500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1049280500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1049280500 Supplier Technidata Supplier No. 1.04928.0500

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
IUPAC Name dipotassium carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color White
pH 11.5 to 12.5 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 500 g
Solubility Information 1120g/L
Percent Purity >99.0%
Grade ACS
Packaging Plastic Bottle
Density 2.43g/3 (20°C)
EINECSNumber 209-529-3