missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate, Anhydrous, Granular, FCC, 99-100.5%, Spectrum™ Chemical
P1237, 584-08-7, K2CO3
$421.50 - $12754.43
Chemical Identifiers
| CAS | 584-08-7 |
|---|---|
| Molecular Formula | CK2O3 |
| Molecular Weight (g/mol) | 138.21 |
| MDL Number | MFCD00011382 |
| InChI Key | BWHMMNNQKKPAPP-UHFFFAOYSA-L |
| IUPAC Name | dipotassium carbonate |
| SMILES | [K+].[K+].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605484
|
Spectrum Chemical Mfg Cor
P1237500GM |
500 g | Poly Bottle |
Each for $421.50
|
|
||||
|
18605485
|
Spectrum Chemical Mfg Cor
P123725KG |
2.5 kg | Poly Pail |
Each for $1,141.54
|
|
||||
|
18605486
|
Spectrum Chemical Mfg Cor
P123712KG |
12 kg | Poly Pail |
Each for $3,339.93
|
|
||||
|
18605487
|
Spectrum Chemical Mfg Cor
P123745KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||