missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, Anhydrous, Granular, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. 18605484
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605484 500 g Poly Bottle
18605485 2.5 kg Poly Pail
18605486 12 kg Poly Pail
18605487 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605484 Supplier Spectrum Chemical Mfg Cor Supplier No. P1237500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P1237, 584-08-7, K2CO3

Spectrum™ Chemical Potassium Carbonate, Anhydrous, Granular, FCC is used as a meat tenderizer, a source of dietary potassium and a buffering agent in wine production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
IUPAC Name dipotassium carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99.0 to 100.5%
Grade FCC
Loss on Drying 0.01
Packaging Poly Bottle