missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, Anhydrous, Granular, FCC, 99-100.5%, Spectrum™ Chemical

P1237, 584-08-7, K2CO3

Supplier:  Spectrum Chemical Mfg Cor P123725KG

 View more versions of this product

Catalog No. 18605485


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Carbonate, Anhydrous, Granular, FCC is used as a meat tenderizer, a source of dietary potassium and a buffering agent in wine production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

584-08-7
2.5 kg
MFCD00011382
[K+].[K+].[O-]C([O-])=O
138.21
FCC
Poly Pail
1
CK2O3
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium carbonate
99.0 to 100.5%
0.01

Chemical Identifiers

584-08-7
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
[K+].[K+].[O-]C([O-])=O
CK2O3
MFCD00011382
dipotassium carbonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions