missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, Certified, 10.0% (w/v) ±0.5% (w/v), LabChem™

Catalog No. LC187824
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC187824 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC187824 Supplier LabChem Supplier No. LC187824

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Color Colorless
CAS Max % 90.64, 9.36
Physical Form Liquid
Quantity 4 L
Linear Formula KBr
Solubility Information Soluble in water
Formula Weight 119
Percent Purity 10.0% ±0.5% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.07g/mL
Decomposition Information Hydrogen bromide
Density 1.07g/mL
Chemical Name or Material Potassium Bromide
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature