missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromate, Granular, ACS, 99.8%, Spectrum™ Chemical
Specifications
Specifications
| pH | 5.0to 9.0 |
| CAS Max % | 1 |
| Physical Form | Granular/Lump |
| Quantity | 45 kg |
| Molecular Weight (g/mol) | 167.00 |
| Percent Purity | 99.8% |
| Grade | ACS/Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Bromate, Granular |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.