missing translation for 'onlineSavingsMsg'
Learn More

Potassium bisulfate, Puriss. p.a., ≥99%, Solstice

Catalog No. 6001194
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001194 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001194 Supplier Solstice Supplier No. 312601KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
IUPAC Name potassium;hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula KHSO4
UN Number 2509
Formula Weight 136.17g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Density 2.32g/mL at 25°C
Chemical Name or Material Potassium Bisulfate
Hazard Statement P261-P280-P305 + P351 + P338-P310
Precautionary Statement H314-H335
EINECSNumber 231-594-1
RTECSNumber TS7200000