missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, 10% (w/v) Aqueous, Ricca Chemical

Catalog No. 5879525
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5879525 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5879525 Supplier Ricca Chemical Company Supplier No. 5879.52.5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium;bromide
SMILES [K+].[Br-]

Specifications

CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Quantity 10 L
Concentration or Composition (by Analyte or Components) 0.100 Normal (N/10)
Packaging Cubitainer
Chemical Name or Material Potassium Biiodate
ShelfLife 18 Months