missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Biiodate, 0.0375 N, Ricca Chemical
Supplier: Ricca Chemical Company R5876000500C
| Quantity | 500 mL |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 13455-24-8 , 7732-18-5 | |
| 389.909 | |
| potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a | |
| potassium;iodic acid;iodate |
| HI2KO6 | |
| ACAYDTMSDROWHW-UHFFFAOYSA-M | |
| 23700942 | |
| OI(=O)=O.[O-]I(=O)=O.[K+] |
Specifications
| Potassium Biiodate | |
| 98.0, 0.12 | |
| HI2KO6 | |
| ACAYDTMSDROWHW-UHFFFAOYSA-M | |
| potassium;iodic acid;iodate | |
| 23700942 | |
| 500 mL |
| 13455-24-8 , 7732-18-5 | |
| 100.0, 0.12 | |
| potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a | |
| OI(=O)=O.[O-]I(=O)=O.[K+] | |
| 389.909 | |
| Amber Glass Bottle | |
| 0.0375Normal |