missing translation for 'onlineSavingsMsg'
Learn More

Potassium Benzoate, NF, 99-101%, Spectrum™ Chemical

Catalog No. 18605449
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605449 500 g Poly Bottle
18605450 2.5 kg Poly Pail
18605451 12 kg Poly Pail
18605452 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605449 Supplier Spectrum Chemical Mfg Cor Supplier No. P1181500GM

P1181, 582-25-2, C7H5KO2

Spectrum™ Chemical Potassium Benzoate, NF is used as a preservative that limits the growth of yeast, mold and some bacteria. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 582-25-2
Molecular Formula C7H5KO2
Molecular Weight (g/mol) 160.21
InChI Key XAEFZNCEHLXOMS-UHFFFAOYSA-M
IUPAC Name potassium benzoate
SMILES [K+].[O-]C(=O)C1=CC=CC=C1

Specifications

Alkalinity To pass test
CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade NF