missing translation for 'onlineSavingsMsg'
Learn More

Potassium Benzoate, Crystal, 99%, Spectrum™ Chemical

$431.64 - $3942.07

Chemical Identifiers

CAS 582-25-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612456
View Documents
Spectrum Chemical Mfg Cor
P1173500GM
500 g Poly Bottle
Each for $431.64
Only null left
 
18612457
View Documents
Spectrum Chemical Mfg Cor
P117325KG
2.5 kg Poly Bottle
Each for $1,329.46
Only null left
 
18612458
View Documents
Spectrum Chemical Mfg Cor
P117312KG
12 kg Poly Pail
Each for $3,942.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

582-25-2

Specifications

582-25-2
500 g
Reagent
1
99%
Poly Bottle
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.