missing translation for 'onlineSavingsMsg'
Learn More

Potassium Benzoate, Crystal, 99%, Spectrum™ Chemical

Catalog No. 18612456
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612456 500 g Poly Bottle
18612457 2.5 kg Poly Bottle
18612458 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612456 Supplier Spectrum Chemical Mfg Cor Supplier No. P1173500GM

Chemical Identifiers

CAS 582-25-2

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.