missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acid Tartrate, Powder, FCC, 99-101%, Spectrum™ Chemical
P1206, 868-14-4, C4H5KO6
$438.46 - $9784.86
Chemical Identifiers
| CAS | 868-14-4 |
|---|---|
| Molecular Formula | C4H5KO6 |
| Molecular Weight (g/mol) | 188.18 |
| MDL Number | MFCD00065392,MFCD00071626,MFCD00064206 |
| InChI Key | KYKNRZGSIGMXFH-ZVGUSBNCSA-M |
| IUPAC Name | potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate |
| SMILES | [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605436
|
Spectrum Chemical Mfg Cor
P1206500GM |
500 g | Poly Bottle |
Each for $438.46
|
|
||||
|
18605437
|
Spectrum Chemical Mfg Cor
P120625KG |
2.5 kg | Poly Pail |
Each for $1,265.36
|
|
||||
|
18605438
|
Spectrum Chemical Mfg Cor
P120612KG |
12 kg | Poly Pail |
Each for $4,669.50
|
|
||||
|
18605439
|
Spectrum Chemical Mfg Cor
P120650KGBL |
50 kg | Fiber Drum |
Each for $9,784.86
|
|
||||
Description
Spectrum™ Chemical Potassium Acid Tartrate, Powder, FCC or potassium bitartrate is called cream of tartar in the food industry. It is useful to stabilize egg whites and whipped cream, prevents sugar syrups from crystallizing and reduces discoloration of boiled vegetables. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 868-14-4 | |
| 188.18 | |
| KYKNRZGSIGMXFH-ZVGUSBNCSA-M | |
| [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O |
| C4H5KO6 | |
| MFCD00065392,MFCD00071626,MFCD00064206 | |
| potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate |
Specifications
| 868-14-4 | |
| 500 g | |
| MFCD00065392,MFCD00071626,MFCD00064206 | |
| [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O | |
| 188.18 | |
| FCC |
| 1 | |
| C4H5KO6 | |
| KYKNRZGSIGMXFH-ZVGUSBNCSA-M | |
| potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate | |
| 99 to 101% | |
| Poly Bottle |