missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acid Tartrate, Powder, FCC, 99-101%, Spectrum™ Chemical

Catalog No. 18605436
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605436 500 g Poly Bottle
18605437 2.5 kg Poly Pail
18605438 12 kg Poly Pail
18605439 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605436 Supplier Spectrum Chemical Mfg Cor Supplier No. P1206500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P1206, 868-14-4, C4H5KO6

Spectrum™ Chemical Potassium Acid Tartrate, Powder, FCC or potassium bitartrate is called cream of tartar in the food industry. It is useful to stabilize egg whites and whipped cream, prevents sugar syrups from crystallizing and reduces discoloration of boiled vegetables. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
IUPAC Name potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99 to 101%
Grade FCC
Packaging Poly Bottle