missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium AA Standard, Certified, 1000ppm K ±10 (1mL = 1mg K), LabChem™
$55.73 - $130.47
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Specifications
| Poly Bottle | |
| 99.81,0.19 | |
| 1g/mL | |
| ClK | |
| KCl | |
| Colorless | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 125 mL | |
| CHEBI:32588 | |
| Certified |
| 7447-40-7,7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm K ±10 (1mL = 1mg K) | |
| MFCD00011360 | |
| Soluble in water | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 | |
| 74.55 | |
| Potassium AA Standard, 1000ppm (1mL = 1mg K) |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature