missing translation for 'onlineSavingsMsg'
Learn More

Potassium AA Standard, Certified, 1000ppm K ±10 (1mL = 1mg K), LabChem™

Supplier:  LabChem LC201007

 View more versions of this product

Catalog No. LC201007


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium AA Standard
Poly Bottle
99.81,0.19
1g/mL
ClK
KCl
Colorless
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
125 mL
CHEBI:32588
Certified
1000ppm (1mL = 1mg K)
7447-40-7,7732-18-5
Passes Test
Liquid
1000 ppm K ±10 (1mL = 1mg K)
MFCD00011360
Soluble in water
[Cl-].[K+]
74.55
4873
74.55

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions