missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium AA Standard, 1000 ppm K in 3% HNO3, Ricca Chemical
$148.37 - $349.17
Chemical Identifiers
| CAS | 7757-79-1 |
|---|---|
| Molecular Formula | KNO3 |
| Molecular Weight (g/mol) | 101.10 |
| MDL Number | MFCD00011409 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| PubChem CID | 24434 |
| ChEBI | CHEBI:63043 |
| IUPAC Name | potassium nitrate |
| SMILES | [K+].[O-][N+]([O-])=O |
Chemical Identifiers
| 7757-79-1 | |
| 101.10 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| CHEBI:63043 | |
| [K+].[O-][N+]([O-])=O |
| KNO3 | |
| MFCD00011409 | |
| 24434 | |
| potassium nitrate |
Specifications
| Natural LDPE Bottle | |
| 94.0,4.22,0.24 | |
| Liquid | |
| MFCD00011409 | |
| < 2 | |
| Standard | |
| [K+].[O-][N+]([O-])=O | |
| 101.10 | |
| 24434 | |
| 1mL = 1mg K (1, 000ppm K)KNO3 in 3% HNO3 |
Safety and Handling
ShelfLife : 24 months