missing translation for 'onlineSavingsMsg'
Learn More

Potassium AA Standard, 1000 ppm K in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company AK1KN500

 View more versions of this product

Catalog No. AK1KN-500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium AA Standard
7757-79-1 , 7697-37-2 , 7757-79-1
97.0,4.39,0.25
KNO3
Colorless
Approximately 0°C
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
500 mL
CHEBI:63043
Natural Poly Bottle
94.0,4.22,0.24
Liquid
MFCD00011409
< 2
Standard
[K+].[O-][N+]([O-])=O
101.10
24434
1mL = 1mg K (1, 000ppm K)KNO3 in 3% HNO3

Chemical Identifiers

7757-79-1
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
[K+].[O-][N+]([O-])=O
KNO3
MFCD00011409
24434
potassium nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents

Promotions

Promotions