missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-methoxypyridine-5-trifluoroborate, 97%, Thermo Scientific™

Catalog No. AAH6164506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6164506 5 g
AAH6164503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6164506 Supplier Thermo Scientific Chemicals Supplier No. H6164506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1144573-61-4
Molecular Formula C6H6BF3KNO
Molecular Weight (g/mol) 215.024
MDL Number MFCD09993111
InChI Key ASWXYOAUUZSFCK-UHFFFAOYSA-N
Synonym potassium 2-methoxypyridine-5-trifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl borate, amtb168, potassium 6-methoxy-3-pyridyltrifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl boranuide
PubChem CID 72698470
IUPAC Name potassium;trifluoro-(6-methoxypyridin-3-yl)boranuide
SMILES [B-](C1=CN=C(C=C1)OC)(F)(F)F.[K+]

Specifications

Quantity 5 g
Formula Weight 215.02
Percent Purity 97%
Chemical Name or Material Potassium 2-methoxypyridine-5-trifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures