missing translation for 'onlineSavingsMsg'
Learn More

Potassium 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide 98.0+%, TCI America™

Catalog No. H10581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10581G 1 g
H10585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10581G Supplier TCI America Supplier No. H10581G

Chemical Identifiers

CAS 588668-97-7
Molecular Formula C3F6KNO4S2
Molecular Weight (g/mol) 331.24
MDL Number MFCD06797108
InChI Key XHXQVKHBZXYEKL-UHFFFAOYSA-N
Synonym 4,4,5,5,6,6-Hexafluoroperhydro-1,3,2-dithiazine 1,1,3,3-Tetraoxide Potassium Salt, 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide Potassium Salt
PubChem CID 44630348
IUPAC Name potassium 4,4,5,5,6,6-hexafluoro-1,1,3,3-tetraoxo-1λ⁶,3λ⁶,2-dithiazinan-2-ide
SMILES [K+].FC1(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C1(F)F

Specifications

Color White
Quantity 1 g
Formula Weight 331.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide