missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ Caprylate Mixed-Mode Cation Exchange Chromatography Resin

Catalog No. A51049
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51049 50 mL
A51050 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A51049 Supplier Thermo Scientific™ Supplier No. A51049

Thermo Scientific™ POROS™ Caprylate Mixed-Mode Chromatography Resin is designed to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode.

POROS Caprylate Mixed-Mode Chromatography Resin is designed to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode. As a mixed-mode, weak cation-exchange resin, POROS Caprylate resin effectively removes other impurities, such as host cell proteins and a wide range of high molecular weight species.

Features of POROS Caprylate Mixed-Mode resin include:

  • Selective capture of aggregates and hydrophobic high-molecular-weight impurities
  • Flow-through design that helps save buffer and material costs
  • Excellent scalability and high productivity
  • Wide operational range
  • Non-animal-derived

POROS Caprylate Mixed-Mode resin operates in flow-through mode which reduces costs, especially when working with more engineered monoclonal antibody derivatives. The unique selectivity of the caprylate ligand contributes to a high yield of the targeted monomer while using established cation exchange resin buffer systems.

Main characteristics

  • Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Average particle size: 50 μm
  • Ligand: octanoic acid
  • Binding capacity: >20 g lysozyme/L resin
  • Typical buffers: typical cation exchange buffers, pH 4.5–7.0, conductivity 10–30 mS/cm
  • Clean-in-place: 0.5 M NaOH
  • Formulation buffer: 0.1 M NaOH
  • Mechanical resistance: 100 bar (10 MPa, 1450 psi)

POROS Caprylate Mixed-Mode resin is manufactured using a synthetic manufacturing process that is free of animal components.

Specifications

Matrix Poly(styrene-co-divinylbenzene)
Shipping Condition Ambient, cold
Particle Size 50 μm
Form Resin Slurry
Product Line POROS
Quantity 50 mL
Target Monoclonal antibodies
Type Bulk Resin
Stationary Phase Caprylic Acid